accessibility menu, dialog, popup

UserName

CAS RN 4743-74-2

H3COHCH2

CAS RN 4743-74-2

IUPAC Name: 2-phenylpent-4-en-2-ol
Synonyms: 2-phenylpent-4-en-2-ol
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: BELRNEPYFJNSPN-UHFFFAOYNA-N
SMILES: CC(O)(CC=C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results