accessibility menu, dialog, popup

UserName

CH3H3C(S)(S)CH3(R)(R)H3C(S)OH(R)H3COH(R)(R)CH3(S)HOO

CAS RN 465-99-6

IUPAC Name: (4aS,6aS,6bR,8aR,9R,10S,12aR,12bR,14bS)-10-hydroxy-9-(hydroxymethyl)-2,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
Synonyms: hederagenin
Molecular Weight (g/mol): 472.71
Molecular Formula: C30H48O4
InChi Key: PGOYMURMZNDHNS-MYPRUECHSA-N
SMILES: CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(O)=O
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results