accessibility menu, dialog, popup

UserName

CAS RN 458-52-6

H3CONH2F

CAS RN 458-52-6

IUPAC Name: 2-fluoro-4-methoxyaniline
Synonyms: 2-fluoro-4-methoxyaniline
Molecular Weight (g/mol): 141.15
Molecular Formula: C7H8FNO
InChi Key: YWUVOJJHVFLNJA-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results