accessibility menu, dialog, popup

UserName

CAS RN 456-71-3

H2NOOFFF

CAS RN 456-71-3

IUPAC Name: 4-(trifluoromethoxy)benzamide
Synonyms: 4-(trifluoromethoxy)benzamide
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: IDIXWLCRJFBQJA-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results