accessibility menu, dialog, popup

UserName

CAS RN 4545-20-4

CH3SOONHN

CAS RN 4545-20-4

IUPAC Name: N'-(diphenylmethylidene)-4-methylbenzene-1-sulfonohydrazide
Synonyms: N'-(diphenylmethylidene)-4-methylbenzenesulfonohydrazide
Molecular Weight (g/mol): 350.44
Molecular Formula: C20H18N2O2S
InChi Key: AVWHDUALELPLAI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NN=C(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results