CAS RN 4431-01-0
CAS RN 4431-01-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ligustilide, MedChemExpress
MedChemExpress Ligustilide is is a bioactive phthalide derivative isolated from Angelica sinensis and Chuanxiong. Ligustilide exhibits neuroprotective, anti-cancer, anti-inflammatory, and vasodilator effects.