accessibility menu, dialog, popup

UserName

CAS RN 4426-76-0

OS

CAS RN 4426-76-0

IUPAC Name: 3,4-dihydro-1H-2-benzothiopyran-4-one
Synonyms: 1,3-dihydro-2-benzothiopyran-4-one
Molecular Weight (g/mol): 164.22
Molecular Formula: C9H8OS
InChi Key: WXGFRIMZEZRKFH-UHFFFAOYSA-N
SMILES: O=C1CSCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results