CAS RN 4374-62-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(Hydroxymethyl)butanoic Acid, TRC
CAS: 4374-62-3 Molecular Formula: C5 H10 O3 Molecular Weight (g/mol): 118.13 Synonym: Butanoic acid, 2-(hydroxymethyl)-,Butyric acid, 2-(hydroxymethyl)- (6CI,8CI),2-(Hydroxymethyl)butanoic acid,2-(Hydroxymethyl)butyric acid,2-Ethyl-3-hydroxypropionic acid,2-Ethylhydracrylic acid,3-Hydroxy-2-ethylpropionic acid,β-Hydroxy-α-ethylpropionic acid IUPAC Name: 2-(hydroxymethyl)butanoic acid SMILES: CCC(CO)C(=O)O