CAS RN 431979-47-4
IUPAC Name:
methyl 2-{2-chloro-6-ethoxy-4-[(3-methyl-5-oxo-1-phenyl-4,5-dihydro-1H-pyrazol-4-ylidene)methyl]phenoxy}acetate
Synonyms:
methyl 2-{2-chloro-6-ethoxy-4-[(3-methyl-5-oxo-1-phenylpyrazol-4-ylidene)methyl]phenoxy}acetate
Molecular Weight (g/mol):
428.87
Molecular Formula:
C22H21ClN2O5
InChi Key:
RKKFQJXGAQWHBZ-UHFFFAOYSA-N
SMILES:
CCOC1=C(OCC(=O)OC)C(Cl)=CC(C=C2C(C)=NN(C2=O)C2=CC=CC=C2)=C1
1
–
1
of
1
results
1
SJ-172550, MedChemExpress
MedChemExpress SJ-172550 is a small molecule inhibitor of MDMX; competes for the wild type p53 peptide binding to MDMX with an EC50 of 5 μM.