CAS RN 43036-06-2
IUPAC Name:
3-(1H-pyrrol-1-yl)propanenitrile
Synonyms:
3-(pyrrol-1-yl)propanenitrile
Molecular Weight (g/mol):
120.16
Molecular Formula:
C7H8N2
InChi Key:
IYOLJLGYJMJLSU-UHFFFAOYSA-N
SMILES:
N#CCCN1C=CC=C1
1
–
1
of
1
results
1
3-(1H-Pyrrol-1-yl)propanenitrile, 97%, Thermo Scientific™
CAS: 43036-06-2 Molecular Formula: C7H8N2 Molecular Weight (g/mol): 120.16 MDL Number: MFCD00003092 InChI Key: IYOLJLGYJMJLSU-UHFFFAOYSA-N Synonym: 3-1h-pyrrol-1-yl propanenitrile,n-2-cyanoethyl pyrrole,1-2-cyanoethyl pyrrole,pyrrole-1-propionitrile,1h-pyrrole-1-propanenitrile,3-pyrrol-1-yl propiononitrile,n-2-cyanoethyl-pyrrole,3-pyrrol-1-yl propanenitrile,n-cyanoethylpyrrole,n-2-cyanoethyl pyrole PubChem CID: 96527 IUPAC Name: 3-pyrrol-1-ylpropanenitrile SMILES: N#CCCN1C=CC=C1