accessibility menu, dialog, popup

UserName

H3CSiCH3H3CNHSiCH3CH3H3CH3CSiCH3H3CNHSiCH3CH3H3CH3CSiCH3H3CNHSiCH3CH3H3CY

CAS RN 41836-28-6

IUPAC Name: tris(bis(trimethylsilyl)amine) yttrium
Synonyms: tris(hexamethyldisilizane) yttrium
Molecular Weight (g/mol): 573.09
Molecular Formula: C18H57N3Si6Y
InChi Key: WJMJCIJUJNENIA-UHFFFAOYSA-N
SMILES: [Y].C[Si](C)(C)N[Si](C)(C)C.C[Si](C)(C)N[Si](C)(C)C.C[Si](C)(C)N[Si](C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)