CAS RN 41717-26-4
IUPAC Name:
1-[(2,4,6-trimethylphenyl)methyl]piperazine
Synonyms:
1-[(2,4,6-trimethylphenyl)methyl]piperazine
Molecular Weight (g/mol):
218.34
Molecular Formula:
C14H22N2
InChi Key:
FOERNUXLFALRDN-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(CN2CCNCC2)C(C)=C1
1
–
1
of
1
results
1
1-(2,4,6-Trimethylbenzyl)piperazine, 97%, Thermo Scientific™
CAS: 41717-26-4 Molecular Formula: C14H22N2 Molecular Weight (g/mol): 218.344 MDL Number: MFCD03094587 InChI Key: FOERNUXLFALRDN-UHFFFAOYSA-N Synonym: 1-2,4,6-trimethylbenzyl piperazine,1-2,4,6-trimethylphenyl methyl piperazine,piperazine, 1-2,4,6-trimethylphenyl methyl,1-2,4,6-trimethylbenzyl-piperazine,1-2,4,6-trimethyl-benzyl-piperazine,1-2 4 6-trimethylbenzyl piperazine PubChem CID: 2737385 IUPAC Name: 1-[(2,4,6-trimethylphenyl)methyl]piperazine SMILES: CC1=CC(=C(C(=C1)C)CN2CCNCC2)C