accessibility menu, dialog, popup

UserName

ClNCOCl

CAS RN 41195-90-8

IUPAC Name: 1,2-dichloro-3-isocyanatobenzene
Synonyms: 1,2-dichloro-3-isocyanatobenzene
Molecular Weight (g/mol): 188.01
Molecular Formula: C7H3Cl2NO
InChi Key: FYWJWWMKCARWQG-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(N=C=O)=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results