accessibility menu, dialog, popup

UserName

CAS RN 406482-19-7

BOH3CFHOOH

CAS RN 406482-19-7

IUPAC Name: (2-fluoro-5-methoxyphenyl)boronic acid
Synonyms: 2-fluoro-5-methoxyphenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: IPTZOWYBCLEBOE-UHFFFAOYSA-N
SMILES: COC1=CC(B(O)O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results