CAS RN 406211-06-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
IKK-IN-1, MedChemExpress
MedChemExpress IKK-IN-1 is an inhibitor of IKK extracted from patent WO2002024679A1, compound example 18-13.