accessibility menu, dialog, popup

UserName

CAS RN 400-72-6

H3COBrFFF

CAS RN 400-72-6

IUPAC Name: 1-bromo-4-methoxy-2-(trifluoromethyl)benzene
Synonyms: 1-bromo-4-methoxy-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 255.03
Molecular Formula: C8H6BrF3O
InChi Key: NSGGOSHNRGWKCM-UHFFFAOYSA-N
SMILES: COC1=CC(=C(Br)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results