CAS RN 398493-79-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PHA-680632, MedChemExpress
MedChemExpress PHA-680632 is an aurora kinase inhibitor with IC50s of 27, 135 and 120 nM for aurora A, B and C, respectively.