accessibility menu, dialog, popup

UserName

CAS RN 39825-33-7

H3CCH3OOCH3NH2

CAS RN 39825-33-7

IUPAC Name: propan-2-yl 2-aminopropanoate
Synonyms: isopropyl 2-aminopropanoate
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: QDQVXVRZVCTVHE-UHFFFAOYNA-N
SMILES: CC(C)OC(=O)C(C)N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)