accessibility menu, dialog, popup

UserName

NH2

CAS RN 3963-62-0

IUPAC Name: 2,2-diphenylethan-1-amine
Synonyms: 2,2-diphenylethanamine
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: RXMTUVIKZRXSSM-UHFFFAOYSA-N
SMILES: NCC(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results