accessibility menu, dialog, popup

UserName

CAS RN 39540-30-2

H3COONH2HCl

CAS RN 39540-30-2

IUPAC Name: octyl 2-aminoacetate hydrochloride
Synonyms: octyl 2-aminoacetate hydrochloride
Molecular Weight (g/mol): 223.74
Molecular Formula: C10H22ClNO2
InChi Key: OORFEZYGSYLFCL-UHFFFAOYSA-N
SMILES: Cl.CCCCCCCCOC(=O)CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)