CAS RN 385380-74-5
CAS RN 385380-74-5
IUPAC Name:
1-(2-fluoro-4-nitrophenyl)pyrrolidine
Synonyms:
1-(2-fluoro-4-nitrophenyl)pyrrolidine
Molecular Weight (g/mol):
210.21
Molecular Formula:
C10H11FN2O2
InChi Key:
SMKXLBISYVJESG-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(F)=C(C=C1)N1CCCC1
1
–
1
of
1
results
1
1-(2-Fluoro-4-nitrophenyl)pyrrolidine, 97%, Thermo Scientific™
CAS: 385380-74-5 Molecular Formula: C10H11FN2O2 Molecular Weight (g/mol): 210.208 InChI Key: SMKXLBISYVJESG-UHFFFAOYSA-N Synonym: 1-2-fluoro-4-nitrophenyl pyrrolidine,3-fluoro-4-pyrrolidin-1-yl nitrobenzene,pyrrolidine, 1-2-fluoro-4-nitrophenyl,pubchem18700,cambridge id 5842403,2-fluoro-4-nitrophenyl pyrrolidine,1-2-fluoro-4-nitro-phenyl pyrrolidine PubChem CID: 2872122 IUPAC Name: 1-(2-fluoro-4-nitrophenyl)pyrrolidine SMILES: C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F