accessibility menu, dialog, popup

UserName

CAS RN 38469-83-9

H3CONNOO

CAS RN 38469-83-9

IUPAC Name: 4-methoxy-2-nitrobenzonitrile
Synonyms: 4-methoxy-2-nitrobenzonitrile
Molecular Weight (g/mol): 178.15
Molecular Formula: C8H6N2O3
InChi Key: UUYPUFCWSUBTFP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C#N)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results