CAS RN 38380-08-4
IUPAC Name:
2,3,3',4,4',5-hexachloro-1,1'-biphenyl
Synonyms:
2,3,3',4,4',5-hexachloro-1,1'-biphenyl
Molecular Weight (g/mol):
360.86
Molecular Formula:
C12H4Cl6
InChi Key:
LCXMEXLGMKFLQO-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1Cl)C1=CC(Cl)=C(Cl)C(Cl)=C1Cl
1
–
1
of
1
results
1
3,4,2',3',4',5'-Hexachlorobiphenyl, TRC
CAS: 38380-08-4 Molecular Formula: C12 H4 Cl6 Molecular Weight (g/mol): 360.88 Synonym: 1,1'-Biphenyl, 2,3,3',4,4',5-hexachloro-,2,3,3',4,4',5-Hexachloro-1,1'-biphenyl,2,3,3',4,4',5-Hexachlorobiphenyl,2,3,3',4,4',5-HxCB,2,3,4,4',5,5'-Hexachlorobiphenyl,2,3,4,5,3',4'-Hexachlorobiphenyl,3,4,2',3',4',5'-Hexachlorobiphenyl,CB 156,PCB 156,2,3,3',4,4',5-HexaCB IUPAC Name: 1,2,3,4-tetrachloro-5-(3,4-dichlorophenyl)benzene SMILES: Clc1ccc(cc1Cl)c2cc(Cl)c(Cl)c(Cl)c2Cl