CAS RN 383145-29-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(4-Fluorophenyl)-1,3-thiazole, TRC
CAS: 383145-29-7 Molecular Formula: C9H6NFS Molecular Weight (g/mol): 179.21 SMILES: Fc1ccc(cc1)c2cscn2