accessibility menu, dialog, popup

UserName

CH3ONN

CAS RN 38028-67-0

IUPAC Name: 3-ethoxypyridazine
Synonyms: pyridazine, 3-ethoxy-
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: ZVVWBPYZGKHHSR-UHFFFAOYSA-N
SMILES: CCOC1=CC=CN=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results