CAS RN 367-33-9
IUPAC Name:
ethyl 2-bromo-4,4,4-trifluorobutanoate
Synonyms:
ethyl 2-bromo-4,4,4-trifluorobutanoate
Molecular Weight (g/mol):
249.03
Molecular Formula:
C6H8BrF3O2
InChi Key:
POVYUFTVLGOXEE-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(Br)CC(F)(F)F
1
–
2
of
2
results
1
Ethyl 2-bromo-4,4,4-trifluorobutyrate, 97%
CAS: 367-33-9 Molecular Formula: C6H8BrF3O2 Molecular Weight (g/mol): 249.027 MDL Number: MFCD10687235 InChI Key: POVYUFTVLGOXEE-UHFFFAOYSA-N Synonym: 2-bromo-4,4,4-trifluorobutyric acid ethyl ester,ethyl 2-bromo-4,4,4-trifluorobutyrate,ethyl 2-bromo-4,4,4-trifluoro-butanoate,butanoic acid,2-bromo-4,4,4-trifluoro-, ethyl ester PubChem CID: 45594306 IUPAC Name: ethyl 2-bromo-4,4,4-trifluorobutanoate SMILES: CCOC(=O)C(CC(F)(F)F)Br