CAS RN 36556-77-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Deethylcyanazine amide, TRC
CAS: 36556-77-1 Molecular Formula: C7H11ClN6O Molecular Weight (g/mol): 230.65 Synonym: 2-[(4-amino-6-chloro-1,3,5-triazin-2-yl)amino]-2-methyl-Propanamide,2-[(4-Amino-6-chloro-1,3,5-triazin-2-yl)amino]-2-methylpropanamide,2-Chloro-4-(1-carbamoyl-1-methylethylamino)-6-amino-1,3,5-triazine,2-Chloro-4-(1-carbamoyl-1-methylethylamino)-6-aminotriazine IUPAC Name: 2-[(4-amino-6-chloro-1,3,5-triazin-2-yl)amino]-2-methylpropanamide SMILES: CC(C)(Nc1nc(N)nc(Cl)n1)C(=O)N