CAS RN 36418-96-9
CAS RN 36418-96-9
IUPAC Name:
3-[3-(benzyloxy)-4-methoxyphenyl]propanoic acid
Synonyms:
3-[3-(benzyloxy)-4-methoxyphenyl]propanoic acid
Molecular Weight (g/mol):
286.33
Molecular Formula:
C17H18O4
InChi Key:
SZORHBHYQVTEEP-UHFFFAOYSA-N
SMILES:
COC1=C(OCC2=CC=CC=C2)C=C(CCC(O)=O)C=C1
1
–
1
of
1
results
1
3-(3-Benzyloxy-4-methoxyphenyl)propionic acid, 96%, Thermo Scientific™
CAS: 36418-96-9 Molecular Formula: C17H18O4 Molecular Weight (g/mol): 286.327 MDL Number: MFCD00227803 InChI Key: SZORHBHYQVTEEP-UHFFFAOYSA-N Synonym: 3-3-benzyloxy-4-methoxyphenyl propanoic acid,3-3-benzyloxy-4-methoxyphenyl propionic acid,3-benzyloxy-4-methoxybenzenepropionic acid PubChem CID: 9971231 IUPAC Name: 3-(4-methoxy-3-phenylmethoxyphenyl)propanoic acid SMILES: COC1=C(C=C(C=C1)CCC(=O)O)OCC2=CC=CC=C2