accessibility menu, dialog, popup

UserName

CAS RN 364-47-6

H3CFFFFFFOH

CAS RN 364-47-6

IUPAC Name: 1-[2,5-bis(trifluoromethyl)phenyl]ethan-1-ol
Synonyms: 1-[2,5-bis(trifluoromethyl)phenyl]ethanol
Molecular Weight (g/mol): 258.16
Molecular Formula: C10H8F6O
InChi Key: JQSGCKQDLFXCFT-UHFFFAOYNA-N
SMILES: CC(O)C1=C(C=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results