accessibility menu, dialog, popup

UserName

CAS RN 3637-63-6

HO

CAS RN 3637-63-6

IUPAC Name: cyclooctylmethanol
Synonyms: cyclooctylmethanol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: ZHPBLHYKDKSZCQ-UHFFFAOYSA-N
SMILES: OCC1CCCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results