accessibility menu, dialog, popup

UserName

ONH2

CAS RN 361393-65-9

IUPAC Name: 1-(2,3-dihydro-1-benzofuran-7-yl)methanamine
Synonyms: 1-(2,3-dihydro-1-benzofuran-7-yl)methanamine
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: LNQLTDVDXCZARO-UHFFFAOYSA-N
SMILES: NCC1=CC=CC2=C1OCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results