CAS RN 361185-42-4
CAS RN 361185-42-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
RBC8, MedChemExpress
MedChemExpress RBC8 is a novel small molecule inhibitor of Ral GTPase; has IC50 of 3.5 μM in H2122 cell and 3.4 μM in H358 cell.
2
Tocris Bioscience™ RBC8
RalA and RalB GTPase inhibitor