accessibility menu, dialog, popup

UserName

CAS RN 36085-73-1

HHClClH2NSNNCH2

CAS RN 36085-73-1

IUPAC Name: dihydrogen 6-(prop-2-en-1-yl)-4H,5H,6H,7H,8H-[1,3]thiazolo[4,5-d]azepin-2-amine dichloride
Synonyms: talipexole
Molecular Weight (g/mol): 282.23
Molecular Formula: C10H17Cl2N3S
InChi Key: DPQAXNSOFFYKDS-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=NC2=C(CCN(CC=C)CC2)S1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results