accessibility menu, dialog, popup

UserName

CAS RN 35789-03-8

H3CSiCH3H3CNHSiCH3CH3H3CH3CSiCH3H3CNHSiCH3CH3H3CH3CSiCH3H3CNHSiCH3CH3H3CGd

CAS RN 35789-03-8

IUPAC Name: tris(bis(trimethylsilyl)amine) gadolinium
Synonyms: tris(hexamethyldisilizane) gadolinium
Molecular Weight (g/mol): 641.44
Molecular Formula: C18H57GdN3Si6
InChi Key: SDYADEGNUOTJQY-UHFFFAOYSA-N
SMILES: [Gd].C[Si](C)(C)N[Si](C)(C)C.C[Si](C)(C)N[Si](C)(C)C.C[Si](C)(C)N[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results