CAS RN 35775-49-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chrysin-7-O-glucuronide, MedChemExpress
MedChemExpress Chrysin-7-O-glucuronide is a flavonoid extracted from Scutellaria baicalensis, with antioxidant activity.