accessibility menu, dialog, popup

UserName

CAS RN 352534-86-2

CH3OClBOHHO

CAS RN 352534-86-2

IUPAC Name: (5-chloro-2-ethoxyphenyl)boronic acid
Synonyms: 5-chloro-2-ethoxyphenylboronic acid
Molecular Weight (g/mol): 200.43
Molecular Formula: C8H10BClO3
InChi Key: OSMBQNBPCSSVMT-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(Cl)C=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results