accessibility menu, dialog, popup

UserName

CAS RN 352534-79-3

HOBOHOF

CAS RN 352534-79-3

IUPAC Name: (2-fluoro-5-formylphenyl)boronic acid
Synonyms: 2-fluoro-5-formylphenylboronic acid
Molecular Weight (g/mol): 167.93
Molecular Formula: C7H6BFO3
InChi Key: SQSWYWCEKCVQEA-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(C=O)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results