accessibility menu, dialog, popup

UserName

CAS RN 352018-97-4

NHNNN

CAS RN 352018-97-4

IUPAC Name: 2-(1,4-diazepan-1-yl)pyridine-3-carbonitrile
Synonyms: 2-(1,4-diazepan-1-yl)pyridine-3-carbonitrile
Molecular Weight (g/mol): 202.26
Molecular Formula: C11H14N4
InChi Key: KQOHMUWBQXSUAK-UHFFFAOYSA-N
SMILES: N#CC1=C(N=CC=C1)N1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results