CAS RN 351003-38-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(Difluoromethoxy)benzenesulfonyl Chloride, TRC
CAS: 351003-38-8 Molecular Formula: C7H5ClF2O3S Molecular Weight (g/mol): 242.63 SMILES: FC(F)Oc1cccc(c1)S(=O)(=O)Cl