accessibility menu, dialog, popup

UserName

CAS RN 351003-22-0

FFFSOOClFFF

CAS RN 351003-22-0

IUPAC Name: 2,5-bis(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 2,5-bis(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 312.61
Molecular Formula: C8H3ClF6O2S
InChi Key: KMEJLLXSUGLEDJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=C(C=C1)C(F)(F)F)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results