CAS RN 350997-12-5
CAS RN 350997-12-5
IUPAC Name:
methyl 2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate
Synonyms:
methyl 2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate
Molecular Weight (g/mol):
251.28
Molecular Formula:
C12H10FNO2S
InChi Key:
DZCBSOKWTGUFCJ-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(N)SC=C1C1=CC=C(F)C=C1
1
–
1
of
1
results
1
Methyl 2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate, 96%, Thermo Scientific™
CAS: 350997-12-5 Molecular Formula: C12H10FNO2S Molecular Weight (g/mol): 251.275 MDL Number: MFCD01827715 InChI Key: DZCBSOKWTGUFCJ-UHFFFAOYSA-N PubChem CID: 712401 IUPAC Name: methyl 2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate SMILES: COC(=O)C1=C(SC=C1C2=CC=C(C=C2)F)N