accessibility menu, dialog, popup

UserName

CAS RN 349094-01-5

H3CCH3CH3NONOOCl

CAS RN 349094-01-5

IUPAC Name: N-benzyl-N-tert-butyl-2-chloro-4-nitrobenzamide
Synonyms: N-benzyl-N-tert-butyl-2-chloro-4-nitrobenzamide
Molecular Weight (g/mol): 346.81
Molecular Formula: C18H19ClN2O3
InChi Key: YIIDQSLHQHNHIJ-UHFFFAOYSA-N
SMILES: CC(C)(C)N(CC1=CC=CC=C1)C(=O)C1=C(Cl)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results