accessibility menu, dialog, popup

UserName

CAS RN 347-63-7

IUPAC Name: 4-(3-fluoro-4-methoxyphenyl)-4-oxobutanoic acid
Synonyms: 4-(3-fluoro-4-methoxyphenyl)-4-oxobutanoic acid
Molecular Weight (g/mol): 226.2
Molecular Formula: C11H11FO4
InChi Key: SNRFVYKMDZSFED-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1F)C(=O)CCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results