CAS RN 34698-85-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Hydroxy DAMPA, TRC
CAS: 34698-85-6 Molecular Formula: C15 H15 N7 O3 Molecular Weight (g/mol): 341.32 Synonym: Benzoic acid, 4-[[(2,4-diamino-7,8-dihydro-7-oxo-6-pteridinyl)methyl]methylamino]-,Benzoic acid, 4-[[(2,4-diamino-1,7-dihydro-7-oxo-6-pteridinyl)methyl]methylamino]- (9CI),4-[[(2,4-Diamino-7,8-dihydro-7-oxo-6-pteridinyl)methyl]methylamino]benzoic acid,4-Amino-4-deoxy-7-hydroxy-N10-methylpteroic acid IUPAC Name: 4-[(2,4-diamino-7-hydroxypteridin-6-yl)methyl-methylamino]benzoic acid SMILES: CN(Cc1nc2c(N)nc(N)nc2nc1O)c3ccc(cc3)C(=O)O