accessibility menu, dialog, popup

UserName

CAS RN 346694-78-8

H3CH3CCH3OONHFFFOOH

CAS RN 346694-78-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-[2-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-[2-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 333.31
Molecular Formula: C15H18F3NO4
InChi Key: XKMOOODKNPYTEE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C(F)(F)F)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)