CAS RN 34404-28-9
CAS RN 34404-28-9
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}pentanedioic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]pentanedioic acid
Molecular Weight (g/mol):
247.25
Molecular Formula:
C10H17NO6
InChi Key:
AQTUACKQXJNHFQ-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CCC(O)=O)C(O)=O
1
–
1
of
1
results
1
N-Boc-D-glutamic acid, 98%
CAS: 34404-28-9 Molecular Formula: C10H17NO6 Molecular Weight (g/mol): 247.247 MDL Number: MFCD00190790 InChI Key: AQTUACKQXJNHFQ-ZCFIWIBFSA-N Synonym: boc-d-glu-oh,boc-d-glutamic acid,n-tert-butoxycarbonyl-d-glutamic acid,r-2-tert-butoxycarbonyl amino pentanedioic acid,d-glutamic acid, n-1,1-dimethylethoxy carbonyl,2r-2-tert-butoxycarbonyl amino pentanedioic acid,pubchem20973,n-boc-d-glutamic acid,ksc493m2t,2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid PubChem CID: 7019046 IUPAC Name: (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid SMILES: CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O