accessibility menu, dialog, popup

UserName

CAS RN 343781-56-6

NBrClOHOCl

CAS RN 343781-56-6

IUPAC Name: 2-bromo-3,5-dichloropyridine-4-carboxylic acid
Synonyms: 2-bromo-3,5-dichloropyridine-4-carboxylic acid
Molecular Weight (g/mol): 270.89
Molecular Formula: C6H2BrCl2NO2
InChi Key: DSKHVZJZZXHWPU-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C(Br)=NC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results