accessibility menu, dialog, popup

UserName

CAS RN 342789-81-5

CH3NNH3CCH3SOOO

CAS RN 342789-81-5

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium methanesulfonate
Synonyms: 1-butyl-3-methylimidazolium mesylate
Molecular Weight (g/mol): 234.31
Molecular Formula: C9H18N2O3S
InChi Key: PUHVBRXUKOGSBC-UHFFFAOYSA-M
SMILES: CS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results