accessibility menu, dialog, popup

UserName

CAS RN 3411-48-1

P

CAS RN 3411-48-1

IUPAC Name: tris(naphthalen-1-yl)phosphane
Synonyms: tris(naphthalen-1-yl)phosphane
Molecular Weight (g/mol): 412.47
Molecular Formula: C30H21P
InChi Key: DMEUUKUNSVFYAA-UHFFFAOYSA-N
SMILES: C1=CC2=CC=CC(P(C3=CC=CC4=CC=CC=C34)C3=C4C=CC=CC4=CC=C3)=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)