accessibility menu, dialog, popup

UserName

CAS RN 332360-01-7

OOHNH2NNH2NH2H2OHCl

CAS RN 332360-01-7

IUPAC Name: 2-amino-5-[(diaminomethylidene)amino]pentanoic acid hydrate hydrochloride
Synonyms: 2-amino-5-[(diaminomethylidene)amino]pentanoic acid hydrate hydrochloride
Molecular Weight (g/mol): 228.68
Molecular Formula: C6H17ClN4O3
InChi Key: NUFCANQNMWVENN-UHFFFAOYNA-N
SMILES: O.Cl.NC(CCCN=C(N)N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)